Search
Search

Gal3st1| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109447.1 | NP_001102917.1 | 3 - 4 | 462 | 106 | |
| XM_006251382.2 | XP_006251444.1 | 2 - 3 | 435 | 106 | ||
| XM_006251383.2 | XP_006251445.1 | 2 - 3 | 2076 | 106 | ||
| XM_008770365.1 | XP_008768587.1 | 1 - 2 | 157 | 106 | ||
| XM_008770366.1 | XP_008768588.1 | 2 - 3 | 538 | 106 | ||
| GenBank mRNA | BC087145.1 | - | 2 - 3 | 212 | 7129361 | 106 |
Set Membership: |
|