Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443252_m1
          See other LETM1 GE Assays ›
          Gene Symbol:
          Letm1
          Gene Name:
          leucine zipper-EF-hand containing transmembrane protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 82227790 - 82267298 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443252_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Letm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005884.1 NP_001005884.1 13 - 14 2273 102
          GenBank mRNA BC083642.1 - 13 - 14 2273 7134917 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000842.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Letm1

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cristae formation
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074260 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074260 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074260 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28136 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13578 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773572 - -
          Affymetrix Rat Gene 1.0 ST Array 10773574 - -
          Affymetrix Rat Gene 1.0 ST Array 10773575 - -
          Affymetrix Rat Gene 1.0 ST Array 10773576 - -
          Affymetrix Rat Gene 1.0 ST Array 10773583 - -
          Affymetrix Rat Gene 1.0 ST Array 10773585 - -
          Affymetrix Rat Gene 1.0 ST Array 10773580 - -
          Affymetrix Rat Gene 1.0 ST Array 10773579 - -
          Affymetrix Rat Gene 1.0 ST Array 10773581 - -
          Affymetrix Rat Gene 1.0 ST Array 10773573 - -
          Affymetrix Rat Gene 1.0 ST Array 10773584 - -
          Affymetrix Rat Gene 1.0 ST Array 10773582 - -
          Affymetrix Rat Gene 1.0 ST Array 10773578 - -
          Affymetrix Rat Gene 1.0 ST Array 10773577 - -
          Affymetrix Rat Gene 1.1 ST Array 10773571 - -
          Rat Genome 230 2.0 Array 1375289_PM_at - -
          Rat Genome 230 2.0 Array 1396013_PM_at - -
          Rat Genome 230 2.0 Array 1371739_PM_at - -
          Rat Genome 230 2.0 Array 1396013_at - -
          Rat Genome 230 2.0 Array 1375289_at - -
          Rat Genome 230 2.0 Array 1371739_at - -
          Affymetrix HT Rat Focus Array 1396013_at - -
          Affymetrix HT Rat Focus Array 1396013_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400005892.rn.1 TC1400000842.rn.1 RTA TC
          Rat Expression Set 230 1396013_at - -
          Rat Expression Set 230 1396013_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10773571 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21819699 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372514 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530368 - -
          RatRef-12 Expression BeadChip ILMN_160584 - -
          RatRef-12 Expression BeadChip ILMN_68647 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline