Search
Search
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191680.1 | NP_001178609.1 | 6 - 7 | 809 | 61 | |
| XM_006251243.2 | XP_006251305.1 | 7 - 8 | 1111 | 61 | ||
| XM_006251244.2 | XP_006251306.1 | 7 - 8 | 1108 | 61 | ||
| XM_006251245.2 | XP_006251307.1 | 7 - 8 | 944 | 61 | ||
| XM_006251246.2 | XP_006251308.1 | 7 - 8 | 941 | 61 | ||
Set Membership: |
|