Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443590_m1
          See other STIM2 GE Assays ›
          Gene Symbol:
          Stim2
          Gene Name:
          stromal interaction molecule 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 59198180 - 59323947 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443590_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stim2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105750.2 NP_001099220.2 3 - 4 610 67
          XM_008770187.1 XP_008768409.1 3 - 4 7252 67
          GenBank mRNA FQ231655.1 - 4 - 5 342 67
          FQ235297.1 - 4 - 5 342 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001868.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          activation of store-operated calcium channel activity
          positive regulation of calcium ion transport
          calcium ion transmembrane transport
          calcium channel regulator activity
          calcium ion binding
          protein binding
          store-operated calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P358664 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38862 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P358664 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P358664 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P331567 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P358664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16328 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80209 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776996 - -
          Affymetrix Rat Gene 1.0 ST Array 10777006 - -
          Affymetrix Rat Gene 1.0 ST Array 10777005 - -
          Affymetrix Rat Gene 1.0 ST Array 10776999 - -
          Affymetrix Rat Gene 1.0 ST Array 10777001 - -
          Affymetrix Rat Gene 1.0 ST Array 10777003 - -
          Affymetrix Rat Gene 1.0 ST Array 10776998 - -
          Affymetrix Rat Gene 1.0 ST Array 10777002 - -
          Affymetrix Rat Gene 1.0 ST Array 10777004 - -
          Affymetrix Rat Gene 1.0 ST Array 10777000 - -
          Affymetrix Rat Gene 1.0 ST Array 10776993 - -
          Affymetrix Rat Gene 1.0 ST Array 10776997 - -
          Affymetrix Rat Gene 1.0 ST Array 10776995 - -
          Affymetrix Rat Gene 1.0 ST Array 10776994 - -
          Affymetrix Rat Gene 1.1 ST Array 10776992 - -
          Rat Genome 230 2.0 Array 1379279_PM_at - -
          Rat Genome 230 2.0 Array 1379279_at - -
          Affymetrix HT Rat Focus Array 1379279_at - -
          Affymetrix HT Rat Focus Array 1379279_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400013488.rn.1 TC1400001868.rn.1 RTA TC
          Rat Expression Set 230 1379279_PM_at - -
          Rat Expression Set 230 1379279_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776992 - -
          Rat Genome U34 Set rc_AI234719_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21406737 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline