Search
Search
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017461.1 | NP_001017461.1 | 2 - 3 | 302 | 76 | |
| XM_006251446.2 | XP_006251508.1 | 2 - 3 | 418 | 76 | ||
| XM_006251447.2 | XP_006251509.1 | 2 - 3 | 564 | 76 | ||
| XM_006251448.2 | XP_006251510.1 | 2 - 3 | 353 | 76 | ||
| GenBank mRNA | BC083811.1 | - | 2 - 3 | 302 | 7110182 | 76 |
| FQ222842.1 | - | 2 - 3 | 277 | 76 | ||
Set Membership: |
|