Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443896_m1
          See other ABCG3L3 GE Assays ›
          Gene Symbol:
          Abcg3l3
          Gene Name:
          ATP-binding cassette, subfamily G (WHITE), member 3-like 3
          Gene Aliases:
          RGD1564709
          Chromosome Location:
          Chr.14: 6058593 - 6104164 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443896_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcg3l3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107205.1 NP_001100675.1 13 - 14 1666 94
          XM_006250606.2 XP_006250668.1 13 - 14 2305 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002461.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P715210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P715210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P715210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P715210 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P715210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771192 - -
          Affymetrix Rat Gene 1.0 ST Array 10771193 - -
          Affymetrix Rat Gene 1.0 ST Array 10775296 - -
          Affymetrix Rat Gene 1.0 ST Array 10771200 - -
          Affymetrix Rat Gene 1.0 ST Array 10775294 - -
          Affymetrix Rat Gene 1.0 ST Array 10771198 - -
          Affymetrix Rat Gene 1.0 ST Array 10771199 - -
          Affymetrix Rat Gene 1.0 ST Array 10775297 - -
          Affymetrix Rat Gene 1.0 ST Array 10771196 - -
          Affymetrix Rat Gene 1.0 ST Array 10771191 - -
          Affymetrix Rat Gene 1.0 ST Array 10771197 - -
          Affymetrix Rat Gene 1.0 ST Array 10775293 - -
          Affymetrix Rat Gene 1.0 ST Array 10771194 - -
          Affymetrix Rat Gene 1.0 ST Array 10775299 - -
          Affymetrix Rat Gene 1.0 ST Array 10771195 - -
          Affymetrix Rat Gene 1.1 ST Array 10771190 - -
          Affymetrix Rat Gene 1.1 ST Array 10775283 - -
          Affymetrix Rat Clariom S Assay PSR1400018256.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018260.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018261.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018255.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018257.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018258.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018259.rn.1 TC1400002461.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10771190 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20732118 - -
          ABI Rat Genome Survey Microarray 21598764 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360371 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline