Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443957_m1
          See other ZDHHC23 GE Assays ›
          Gene Symbol:
          Zdhhc23
          Gene Name:
          zinc finger, DHHC-type containing 23
          Gene Aliases:
          DHHC23, nidd
          Chromosome Location:
          Chr.11: 61661086 - 61677547 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443957_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_213627.2 NP_998792.2 1 - 2 261 122
          XM_008768738.1 XP_008766960.1 1 - 2 281 122
          XM_008768739.1 XP_008766961.1 1 - 2 281 122
          XM_008768740.1 XP_008766962.1 2 - 3 523 122
          XM_008768741.1 XP_008766963.1 1 - 2 281 122
          XM_008768742.1 XP_008766964.1 1 - 2 281 122
          GenBank mRNA AB098078.1 - 2 - 3 299 122
          AY886538.1 - 1 - 2 261 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000611.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zdhhc23

          Gene Ontology Categories:

          Function(s) Process(es)

          protein palmitoylation
          protein localization to plasma membrane
          zinc ion binding
          protein-cysteine S-palmitoyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P456599 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160556 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456599 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160556 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160556 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P456599 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P456599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64363 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34839 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751191 - -
          Affymetrix Rat Gene 1.0 ST Array 10751193 - -
          Affymetrix Rat Gene 1.0 ST Array 10751194 - -
          Affymetrix Rat Gene 1.0 ST Array 10751192 - -
          Affymetrix Rat Gene 1.0 ST Array 10751195 - -
          Affymetrix Rat Gene 1.1 ST Array 10751190 - -
          Rat Genome 230 2.0 Array 1390607_PM_at - -
          Rat Genome 230 2.0 Array 1390607_at - -
          Affymetrix HT Rat Focus Array 1390607_PM_at - -
          Affymetrix HT Rat Focus Array 1390607_at - -
          Affymetrix Rat Clariom S Assay PSR1100003248.rn.1 TC1100000611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003253.rn.1 TC1100000611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003250.rn.1 TC1100000611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003249.rn.1 TC1100000611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003254.rn.1 TC1100000611.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003252.rn.1 TC1100000611.rn.1 RTA TC
          Rat Expression Set 230 1390607_PM_at - -
          Rat Expression Set 230 1390607_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751190 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22143516 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366047 - -
          RatRef-12 Expression BeadChip ILMN_55318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline