Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01443979_m1
          See other CD96 GE Assays ›
          Gene Symbol:
          Cd96
          Gene Name:
          CD96 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 57108761 - 57183855 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01443979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025032.1 NP_001020203.1 14 - 15 1733 72
          XM_008768647.1 XP_008766869.1 14 - 15 1937 72
          GenBank mRNA BC091206.1 - 14 - 15 1733 7380788 72
          FQ228851.1 - 14 - 15 1721 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000563.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd96

          Panther Classification:

          Molecular Function -

          defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          cell adhesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P150367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P150367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P150367 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P150367 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P150367 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36765 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55101 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751003 - -
          Affymetrix Rat Gene 1.0 ST Array 10751011 - -
          Affymetrix Rat Gene 1.0 ST Array 10751008 - -
          Affymetrix Rat Gene 1.0 ST Array 10751000 - -
          Affymetrix Rat Gene 1.0 ST Array 10751009 - -
          Affymetrix Rat Gene 1.0 ST Array 10750997 - -
          Affymetrix Rat Gene 1.0 ST Array 10751005 - -
          Affymetrix Rat Gene 1.0 ST Array 10751006 - -
          Affymetrix Rat Gene 1.0 ST Array 10751010 - -
          Affymetrix Rat Gene 1.0 ST Array 10750999 - -
          Affymetrix Rat Gene 1.0 ST Array 10751001 - -
          Affymetrix Rat Gene 1.0 ST Array 10750998 - -
          Affymetrix Rat Gene 1.0 ST Array 10751004 - -
          Affymetrix Rat Gene 1.0 ST Array 10751002 - -
          Affymetrix Rat Gene 1.0 ST Array 10751007 - -
          Affymetrix Rat Gene 1.1 ST Array 10750996 - -
          Rat Genome 230 2.0 Array 1382507_PM_at - -
          Rat Genome 230 2.0 Array 1382507_at - -
          Affymetrix HT Rat Focus Array 1382507_PM_at - -
          Affymetrix HT Rat Focus Array 1382507_at - -
          Affymetrix Rat Clariom S Assay PSR1100002858.rn.1 TC1100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002857.rn.1 TC1100000563.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100002859.rn.1 TC1100000563.rn.1 RTA TC
          Rat Expression Set 230 1382507_at - -
          Rat Expression Set 230 1382507_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750996 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20872937 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353555 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline