Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444029_m1
          See other HGD GE Assays ›
          Gene Symbol:
          Hgd
          Gene Name:
          homogentisate 1, 2-dioxygenase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 65983221 - 66034555 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444029_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hgd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012145.1 NP_001012145.1 13 - 14 1328 69
          GenBank mRNA BC078948.1 - 13 - 14 1328 7109741 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001669.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hgd

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular amino acid metabolic process
          L-phenylalanine catabolic process
          tyrosine metabolic process
          biological_process
          oxidation-reduction process
          molecular_function
          homogentisate 1,2-dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163627 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033795 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033795 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P338149 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20145 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754244 - -
          Affymetrix Rat Gene 1.0 ST Array 10754241 - -
          Affymetrix Rat Gene 1.0 ST Array 10754250 - -
          Affymetrix Rat Gene 1.0 ST Array 10754245 - -
          Affymetrix Rat Gene 1.0 ST Array 10754243 - -
          Affymetrix Rat Gene 1.0 ST Array 10754237 - -
          Affymetrix Rat Gene 1.0 ST Array 10754236 - -
          Affymetrix Rat Gene 1.0 ST Array 10754242 - -
          Affymetrix Rat Gene 1.0 ST Array 10754249 - -
          Affymetrix Rat Gene 1.0 ST Array 10754239 - -
          Affymetrix Rat Gene 1.0 ST Array 10754247 - -
          Affymetrix Rat Gene 1.0 ST Array 10754248 - -
          Affymetrix Rat Gene 1.0 ST Array 10754246 - -
          Affymetrix Rat Gene 1.0 ST Array 10754238 - -
          Affymetrix Rat Gene 1.0 ST Array 10754240 - -
          Affymetrix Rat Gene 1.1 ST Array 10754235 - -
          Rat Genome 230 2.0 Array 1398514_at - -
          Rat Genome 230 2.0 Array 1398514_PM_at - -
          Affymetrix HT Rat Focus Array 1398514_PM_at - -
          Affymetrix HT Rat Focus Array 1398514_at - -
          Affymetrix Rat Clariom S Assay PSR1100010115.rn.1 TC1100001669.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100010116.rn.1 TC1100001669.rn.1 RTA TC
          Rat Expression Set 230 1398514_PM_at - -
          Rat Expression Set 230 1398514_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754235 - -
          Rat Genome U34 Set rc_AI232328_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22067251 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376689 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline