Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444108_m1
          See other GSK3B GE Assays ›
          Gene Symbol:
          Gsk3b
          Gene Name:
          glycogen synthase kinase 3 beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 65060884 - 65209543 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444108_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsk3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032080.1 NP_114469.1 2 - 3 421 96
          XM_006248373.2 XP_006248435.1 2 - 3 1127 96
          XM_006248374.2 XP_006248436.1 2 - 3 1127 96
          XM_006248375.2 XP_006248437.1 2 - 3 1127 96
          XM_008768745.1 XP_008766967.1 2 - 3 1127 96
          GenBank mRNA X53428.1 - 2 - 3 396 96
          X73653.1 - 2 - 3 421 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001658.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gsk3b

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          re-entry into mitotic cell cycle
          epithelial to mesenchymal transition
          positive regulation of cell-matrix adhesion
          glycogen metabolic process
          regulation of gene expression by genetic imprinting
          protein phosphorylation
          protein export from nucleus
          ER overload response
          establishment or maintenance of cell polarity
          axonogenesis
          myoblast fusion
          circadian rhythm
          organ morphogenesis
          response to lithium ion
          negative regulation of cardiac muscle hypertrophy
          positive regulation of peptidyl-threonine phosphorylation
          regulation of neuron projection development
          negative regulation of neuron projection development
          negative regulation of neuron maturation
          myotube differentiation
          Wnt signaling pathway
          phosphorylation
          cell migration
          peptidyl-serine phosphorylation
          hippocampus development
          establishment of cell polarity
          negative regulation of protein complex assembly
          positive regulation of protein complex assembly
          negative regulation of protein binding
          positive regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of microtubule-based process
          positive regulation of peptidyl-serine phosphorylation
          protein localization to microtubule
          intracellular signal transduction
          cellular response to interleukin-3
          response to drug
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAP kinase activity
          hypermethylation of CpG island
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          fat cell differentiation
          positive regulation of axon extension
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          positive regulation of protein export from nucleus
          regulation of neuronal synaptic plasticity
          negative regulation of dendrite morphogenesis
          negative regulation of NFAT protein import into nucleus
          canonical Wnt signaling pathway
          superior temporal gyrus development
          cellular response to mechanical stimulus
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          positive regulation of stem cell differentiation
          RNA polymerase II transcription factor binding
          p53 binding
          protein kinase activity
          protein serine/threonine kinase activity
          integrin binding
          protein binding
          ATP binding
          beta-catenin binding
          transcription factor binding
          kinase activity
          protein kinase binding
          ubiquitin protein ligase binding
          protein kinase A catalytic subunit binding
          ionotropic glutamate receptor binding
          tau protein binding
          tau-protein kinase activity
          NF-kappaB binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P620042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52776 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P620042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P620042 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P620042 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P620042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19146 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40756 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754196 - -
          Affymetrix Rat Gene 1.0 ST Array 10754195 - -
          Affymetrix Rat Gene 1.0 ST Array 10754200 - -
          Affymetrix Rat Gene 1.0 ST Array 10754198 - -
          Affymetrix Rat Gene 1.0 ST Array 10754194 - -
          Affymetrix Rat Gene 1.0 ST Array 10754199 - -
          Affymetrix Rat Gene 1.0 ST Array 10754191 - -
          Affymetrix Rat Gene 1.0 ST Array 10754197 - -
          Affymetrix Rat Gene 1.0 ST Array 10754193 - -
          Affymetrix Rat Gene 1.0 ST Array 10754189 - -
          Affymetrix Rat Gene 1.0 ST Array 10754190 - -
          Affymetrix Rat Gene 1.1 ST Array 10754188 - -
          Rat Genome 230 2.0 Array 1370267_PM_at - -
          Rat Genome 230 2.0 Array 1370267_at - -
          Affymetrix HT Rat Focus Array 1370267_at - -
          Affymetrix HT Rat Focus Array 1370267_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100010044.rn.1 TC1100001658.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100010049.rn.1 TC1100001658.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100010043.rn.1 TC1100001658.rn.1 RTA TC
          Rat Expression Set 230 1370267_PM_at - -
          Rat Expression Set 230 1370267_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754188 - -
          Rat Genome U34 Set X53428cds_s_at - -
          Rat Genome U34 Set X73653_at - -
          Rat Genome U34 Set X53428cds_s_at - -
          Rat Genome U34 Set X73653_at - -
          Rat Genome U34 Set X53428cds_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349648 - -
          RatRef-12 Expression BeadChip ILMN_49666 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline