Search
Search

RGD1560175| Gene Symbol: RGD1560175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248356.2 | XP_006248418.1 | 6 - 7 | 658 | 99 | |
| XM_006248357.2 | XP_006248419.1 | 5 - 6 | 542 | 99 | ||
| XM_221454.8 | XP_221454.5 | 6 - 7 | 793 | 99 | ||
Set Membership: |
|