Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444326_m1
          See other TMEM30C GE Assays ›
          Gene Symbol:
          Tmem30c
          Gene Name:
          transmembrane protein 30C
          Gene Aliases:
          RGD1306090
          Chromosome Location:
          Chr.11: 45337518 - 45357434 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444326_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem30c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105889.1 NP_001099359.1 7 - 8 938 61
          XM_008768632.1 XP_008766854.1 7 - 8 1175 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000456.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P473763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473763 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P473763 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750708 - -
          Affymetrix Rat Gene 1.0 ST Array 10750704 - -
          Affymetrix Rat Gene 1.0 ST Array 10750707 - -
          Affymetrix Rat Gene 1.0 ST Array 10750702 - -
          Affymetrix Rat Gene 1.0 ST Array 10750703 - -
          Affymetrix Rat Gene 1.0 ST Array 10750706 - -
          Affymetrix Rat Gene 1.0 ST Array 10750705 - -
          Affymetrix Rat Gene 1.1 ST Array 10750701 - -
          Affymetrix Rat Clariom S Assay PSR1100002208.rn.1 TC1100000456.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10750701 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21182356 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline