Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444501_m1
          See other CHUK GE Assays ›
          Gene Symbol:
          Chuk
          Gene Name:
          conserved helix-loop-helix ubiquitous kinase
          Gene Aliases:
          Ikbka, Ikka
          Chromosome Location:
          Chr.1: 263848829 - 263884354 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444501_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chuk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107588.1 NP_001101058.1 2 - 3 303 69
          XM_006231481.2 XP_006231543.1 2 - 3 300 69
          XM_008760418.1 XP_008758640.1 2 - 3 301 69
          XM_008760419.1 XP_008758641.1 2 - 3 301 69
          GenBank mRNA BC168905.1 - 2 - 3 278 9088878 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007759.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelial sheet
          skeletal muscle contraction
          protein phosphorylation
          I-kappaB phosphorylation
          Rho protein signal transduction
          lactation
          response to toxic substance
          response to organic substance
          response to acetate
          osteoclast differentiation
          response to lipopolysaccharide
          response to hydroperoxide
          mammary gland epithelial cell proliferation
          response to muscle stretch
          odontogenesis of dentin-containing tooth
          response to drug
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to amino acid
          positive regulation of transcription from RNA polymerase II promoter
          striated muscle cell differentiation
          mammary gland alveolus development
          cellular response to tumor necrosis factor
          protein kinase activity
          ATP binding
          IkappaB kinase activity
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P269609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269609 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P269609 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715516 - -
          Affymetrix Rat Gene 1.0 ST Array 10715499 - -
          Affymetrix Rat Gene 1.0 ST Array 10715504 - -
          Affymetrix Rat Gene 1.0 ST Array 10715505 - -
          Affymetrix Rat Gene 1.0 ST Array 10715507 - -
          Affymetrix Rat Gene 1.0 ST Array 10715508 - -
          Affymetrix Rat Gene 1.0 ST Array 10715510 - -
          Affymetrix Rat Gene 1.0 ST Array 10715502 - -
          Affymetrix Rat Gene 1.0 ST Array 10715503 - -
          Affymetrix Rat Gene 1.0 ST Array 10715517 - -
          Affymetrix Rat Gene 1.0 ST Array 10715515 - -
          Affymetrix Rat Gene 1.0 ST Array 10715500 - -
          Affymetrix Rat Gene 1.0 ST Array 10715512 - -
          Affymetrix Rat Gene 1.0 ST Array 10715514 - -
          Affymetrix Rat Gene 1.0 ST Array 10715506 - -
          Affymetrix Rat Gene 1.0 ST Array 10715501 - -
          Affymetrix Rat Gene 1.0 ST Array 10715511 - -
          Affymetrix Rat Gene 1.0 ST Array 10715509 - -
          Affymetrix Rat Gene 1.0 ST Array 10715518 - -
          Affymetrix Rat Gene 1.0 ST Array 10715513 - -
          Affymetrix Rat Gene 1.0 ST Array 10715498 - -
          Affymetrix Rat Gene 1.1 ST Array 10715497 - -
          Rat Genome 230 2.0 Array 1373517_at - -
          Rat Genome 230 2.0 Array 1373517_PM_at - -
          Affymetrix HT Rat Focus Array 1373517_at - -
          Affymetrix HT Rat Focus Array 1373517_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100062510.rn.1 TC0100007759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062507.rn.1 TC0100007759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062508.rn.1 TC0100007759.rn.1 RTA TC
          Rat Expression Set 230 1373517_at - -
          Rat Expression Set 230 1373517_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715497 - -
          Rat Genome U34 Set rc_AI234361_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20731179 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline