Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444586_m1
          See other RAB11FIP2 GE Assays ›
          Gene Symbol:
          Rab11fip2
          Gene Name:
          RAB11 family interacting protein 2 (class I)
          Gene Aliases:
          RGD1308538
          Chromosome Location:
          Chr.1: 281065346 - 281101163 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01444586_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab11fip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107447.2 NP_001100917.2 4 - 5 1315 110
          GenBank mRNA BC098050.1 - 1353 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007980.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          establishment of cell polarity
          insulin secretion involved in cellular response to glucose stimulus
          regulated exocytosis
          molecular_function
          Rab GTPase binding
          protein kinase binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P515679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177411 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P515679 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P515679 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P515679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515679 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P418753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731145 - -
          Affymetrix Rat Gene 1.0 ST Array 10731142 - -
          Affymetrix Rat Gene 1.0 ST Array 10731146 - -
          Affymetrix Rat Gene 1.0 ST Array 10731143 - -
          Affymetrix Rat Gene 1.0 ST Array 10731144 - -
          Affymetrix Rat Gene 1.1 ST Array 10731140 - -
          Rat Genome 230 2.0 Array 1394607_PM_at - -
          Rat Genome 230 2.0 Array 1394607_at - -
          Affymetrix HT Rat Focus Array 1394607_at - -
          Affymetrix HT Rat Focus Array 1394607_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100064266.rn.1 TC0100007980.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100064265.rn.1 TC0100007980.rn.1 RTA TC
          Rat Expression Set 230 1394607_PM_at - -
          Rat Expression Set 230 1394607_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731140 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20740998 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0