Search
Search
| Gene Symbol: Xrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243594.2 | XP_006243656.1 | 16 - 17 | 2913 | 71 | |
| XM_006243596.2 | XP_006243658.1 | 16 - 17 | 2913 | 71 | ||
| XM_006243597.2 | XP_006243659.1 | 16 - 17 | 2913 | 71 | ||
| XM_006243598.2 | XP_006243660.1 | 16 - 17 | 2913 | 71 | ||
| XM_006243599.2 | XP_006243661.1 | 16 - 17 | 2913 | 71 | ||
| XM_008766573.1 | XP_008764795.1 | 16 - 17 | 2913 | 71 | ||
| XM_217233.8 | XP_217233.5 | 16 - 17 | 2913 | 71 | ||
Set Membership: |
|