Search
Search
| Gene Symbol: Celsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031320.1 | NP_112610.1 | 1 - 2 | 4006 | 63 | |
| XM_006243860.2 | XP_006243922.1 | 1 - 2 | 3743 | 63 | ||
| XM_006243861.2 | XP_006243923.1 | 1 - 2 | 3743 | 63 | ||
| GenBank mRNA | AB011528.1 | - | 1 - 2 | 4006 | 63 | |
Set Membership: |
|