Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01445298_m1
          See other RASSF1 GE Assays ›
          Gene Symbol:
          Rassf1
          Gene Name:
          Ras association (RalGDS/AF-6) domain family member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 116307149 - 116318289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01445298_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rassf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007754.1 NP_001007755.1 4 - 5 705 71
          NM_001037555.1 NP_001032644.1 5 - 6 897 71
          GenBank mRNA AB202124.1 - 5 - 6 897 71
          BC079237.1 - 4 - 5 705 7132629 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001507.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rassf1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to DNA damage stimulus
          cell cycle arrest
          signal transduction
          Ras protein signal transduction
          biological_process
          positive regulation of protein ubiquitination
          protein stabilization
          negative regulation of cell cycle arrest
          protein binding
          identical protein binding
          metal ion binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P173575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P173575 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P173575 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P173575 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68036 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912997 - -
          Affymetrix Rat Gene 1.0 ST Array 10912995 - -
          Affymetrix Rat Gene 1.0 ST Array 10912992 - -
          Affymetrix Rat Gene 1.0 ST Array 10912996 - -
          Affymetrix Rat Gene 1.0 ST Array 10912994 - -
          Affymetrix Rat Gene 1.0 ST Array 10912991 - -
          Affymetrix Rat Gene 1.0 ST Array 10912998 - -
          Affymetrix Rat Gene 1.0 ST Array 10912993 - -
          Affymetrix Rat Gene 1.1 ST Array 10912990 - -
          Rat Genome 230 2.0 Array 1382449_PM_at - -
          Rat Genome 230 2.0 Array 1392688_at - -
          Rat Genome 230 2.0 Array 1373989_at - -
          Rat Genome 230 2.0 Array 1373989_PM_at - -
          Rat Genome 230 2.0 Array 1392688_PM_at - -
          Rat Genome 230 2.0 Array 1382449_at - -
          Affymetrix HT Rat Focus Array 1373989_PM_at - -
          Affymetrix HT Rat Focus Array 1373989_at - -
          Affymetrix HT Rat Focus Array 1382449_PM_at - -
          Affymetrix HT Rat Focus Array 1382449_at - -
          Affymetrix HT Rat Focus Array 1392688_at - -
          Affymetrix HT Rat Focus Array 1392688_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800011082.rn.1 TC0800001507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800011081.rn.1 TC0800001507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800011083.rn.1 TC0800001507.rn.1 RTA TC
          Rat Expression Set 230 1373989_at - -
          Rat Expression Set 230 1373989_PM_at - -
          Rat Expression Set 230 1392688_PM_at - -
          Rat Expression Set 230 1392688_at - -
          Rat Expression Set 230 1382449_PM_at - -
          Rat Expression Set 230 1382449_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912990 - -
          Rat Genome U34 Set rc_AI230845_at - -
          Rat Genome U34 Set rc_AI113186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21467052 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354445 - -
          RatRef-12 Expression BeadChip ILMN_62008 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline