Search
Search
| Gene Symbol: Nicn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034999.1 | NP_001030171.1 | 1 - 2 | 207 | 91 | |
| XM_006243854.2 | XP_006243916.1 | 1 - 2 | 291 | 91 | ||
| GenBank mRNA | BC101861.1 | - | 1 - 2 | 207 | 7936966 | 91 |
Set Membership: |
|