Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01446135_m1
          See other AZI2 GE Assays ›
          Gene Symbol:
          Azi2
          Gene Name:
          5-azacytidine induced 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 126411044 - 126435581 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01446135_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Azi2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025705.1 NP_001020876.1 5 - 6 962 82
          XM_006244032.2 XP_006244094.1 6 - 7 1833 82
          GenBank mRNA BC098680.1 - 5 - 6 962 7384893 82
          FQ221679.1 - 6 - 7 1082 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001703.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Azi2

          Gene Ontology Categories:

          Function(s)

          mitotic cell cycle
          cytokine production
          I-kappaB kinase/NF-kappaB signaling
          interferon-alpha production
          interferon-gamma production
          interleukin-6 production
          tumor necrosis factor production
          T cell activation
          dendritic cell proliferation
          dendritic cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14849 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14849 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14849 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P291114 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14849 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1040055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914077 - -
          Affymetrix Rat Gene 1.0 ST Array 10914082 - -
          Affymetrix Rat Gene 1.0 ST Array 10914075 - -
          Affymetrix Rat Gene 1.0 ST Array 10914076 - -
          Affymetrix Rat Gene 1.0 ST Array 10914078 - -
          Affymetrix Rat Gene 1.0 ST Array 10914080 - -
          Affymetrix Rat Gene 1.0 ST Array 10914079 - -
          Affymetrix Rat Gene 1.0 ST Array 10914081 - -
          Affymetrix Rat Gene 1.1 ST Array 10914074 - -
          Rat Genome 230 2.0 Array 1388631_at - -
          Rat Genome 230 2.0 Array 1388631_PM_at - -
          Rat Genome 230 2.0 Array 1394710_at - -
          Rat Genome 230 2.0 Array 1394710_PM_at - -
          Affymetrix HT Rat Focus Array 1394710_PM_at - -
          Affymetrix HT Rat Focus Array 1394710_at - -
          Affymetrix HT Rat Focus Array 1388631_PM_at - -
          Affymetrix HT Rat Focus Array 1388631_at - -
          Affymetrix Rat Clariom S Assay PSR0800013407.rn.1 TC0800001703.rn.1 RTA TC
          Rat Expression Set 230 1388631_PM_at - -
          Rat Expression Set 230 1388631_at - -
          Rat Expression Set 230 1394710_at - -
          Rat Expression Set 230 1394710_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10914074 - -
          Rat Genome U34 Set rc_AI177093_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20948237 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline