Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01446616_m1
See other LOC498675 GE Assays ›
Gene Symbol:
LOC498675
Gene Name:
hypothetical LOC498675
Gene Aliases:
-
Chromosome Location:
Chr.8: 117170620 - 117175235 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01446616_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LOC498675
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001109113.1
NP_001102583.1
2 - 3
423
78
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0800001542.rn.1
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Rattus norvegicus
Probe spans exons
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0800011502.rn.1
TC0800001542.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0800011504.rn.1
TC0800001542.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0800011503.rn.1
TC0800001542.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline