Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01446631_m1
          See other ELOVL5 GE Assays ›
          Gene Symbol:
          Elovl5
          Gene Name:
          ELOVL fatty acid elongase 5
          Gene Aliases:
          rELO1
          Chromosome Location:
          Chr.8: 85220947 - 85287449 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01446631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elovl5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134382.1 NP_599209.1 6 - 7 762 81
          XM_006243417.1 XP_006243479.1 7 - 8 927 81
          GenBank mRNA AB071985.1 - 6 - 7 762 81
          HQ404314.1 - 6 - 7 762 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001151.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Elovl5

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          unsaturated fatty acid biosynthetic process
          fatty acid elongation, monounsaturated fatty acid
          fatty acid elongation, polyunsaturated fatty acid
          very long-chain fatty acid biosynthetic process
          fatty acid elongase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170434 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128862 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128862 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128862 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53800 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911772 - -
          Affymetrix Rat Gene 1.0 ST Array 10911766 - -
          Affymetrix Rat Gene 1.0 ST Array 10911770 - -
          Affymetrix Rat Gene 1.0 ST Array 10911767 - -
          Affymetrix Rat Gene 1.0 ST Array 10911769 - -
          Affymetrix Rat Gene 1.0 ST Array 10911768 - -
          Affymetrix Rat Gene 1.0 ST Array 10911771 - -
          Affymetrix Rat Gene 1.1 ST Array 10911765 - -
          Rat Genome 230 2.0 Array 1387630_PM_at - -
          Rat Genome 230 2.0 Array 1388348_PM_at - -
          Rat Genome 230 2.0 Array 1387630_at - -
          Rat Genome 230 2.0 Array 1388348_at - -
          Affymetrix HT Rat Focus Array 1387630_at - -
          Affymetrix HT Rat Focus Array 1387630_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800008431.rn.1 TC0800001151.rn.1 RTA TC
          Rat Expression Set 230 1388348_PM_at - -
          Rat Expression Set 230 1387630_at - -
          Rat Expression Set 230 1387630_PM_at - -
          Rat Expression Set 230 1388348_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911765 - -
          Rat Genome U34 Set rc_AA892832_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21660819 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349146 - -
          RatRef-12 Expression BeadChip ILMN_56318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline