Search
Search
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109296.1 | NP_001102766.1 | 7 - 8 | 867 | 69 | |
| XM_008766561.1 | XP_008764783.1 | 8 - 9 | 1327 | 69 | ||
| XM_008766562.1 | XP_008764784.1 | 6 - 7 | 744 | 69 | ||
| GenBank mRNA | FQ233502.1 | - | 8 - 9 | 931 | 69 | |
Set Membership: |
|