Search
Search

Abhd14a| Gene Symbol: Abhd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009670.1 | NP_001009670.1 | 4 - 5 | 829 | 94 | |
| XM_006243717.2 | XP_006243779.1 | 3 - 4 | 670 | 94 | ||
| XM_006243720.2 | XP_006243782.1 | 3 - 4 | 1424 | 94 | ||
| XM_008766494.1 | XP_008764716.1 | 4 - 5 | 1241 | 94 | ||
| XM_008766495.1 | XP_008764717.1 | 5 - 6 | 895 | 94 | ||
| XM_008766496.1 | XP_008764718.1 | 4 - 5 | 844 | 94 | ||
| XM_008766497.1 | XP_008764719.1 | 4 - 5 | 713 | 94 | ||
| XM_008766498.1 | XP_008764720.1 | 3 - 4 | 665 | 94 | ||
| XM_008766499.1 | XP_008764721.1 | 3 - 4 | 670 | 94 | ||
| GenBank mRNA | BC088472.1 | - | 4 - 5 | 829 | 7128506 | 94 |
Set Membership: |
|