Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447402_m1
          See other FILIP1 GE Assays ›
          Gene Symbol:
          Filip1
          Gene Name:
          filamin A interacting protein 1
          Gene Aliases:
          Filip
          Chromosome Location:
          Chr.8: 87268176 - 87419370 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447402_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Filip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145682.1 NP_663715.1 5 - 6 3509 65
          GenBank mRNA AB055759.1 - 5 - 6 3509 65
          D87257.1 - 2 - 3 2930 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003006.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Filip1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P596942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162168 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P596942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P596942 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P596942 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P596942 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000722 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918959 - -
          Affymetrix Rat Gene 1.0 ST Array 10918957 - -
          Affymetrix Rat Gene 1.0 ST Array 10918958 - -
          Affymetrix Rat Gene 1.0 ST Array 10918956 - -
          Affymetrix Rat Gene 1.1 ST Array 10918955 - -
          Rat Genome 230 2.0 Array 1375089_PM_at - -
          Rat Genome 230 2.0 Array 1379463_at - -
          Rat Genome 230 2.0 Array 1375089_at - -
          Rat Genome 230 2.0 Array 1379463_PM_at - -
          Affymetrix HT Rat Focus Array 1379463_at - -
          Affymetrix HT Rat Focus Array 1379463_PM_at - -
          Affymetrix HT Rat Focus Array 1375089_at - -
          Affymetrix HT Rat Focus Array 1375089_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800024087.rn.1 TC0800003006.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800024088.rn.1 TC0800003006.rn.1 RTA TC
          Rat Expression Set 230 1375089_at - -
          Rat Expression Set 230 1375089_PM_at - -
          Rat Expression Set 230 1379463_PM_at - -
          Rat Expression Set 230 1379463_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918955 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21849570 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376850 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline