Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447445_m1
          See other CLDN18 GE Assays ›
          Gene Symbol:
          Cldn18
          Gene Name:
          claudin 18
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 107933730 - 107960664 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014096.1 NP_001014118.1 3 - 4 537 73
          XM_006243629.2 XP_006243691.1 3 - 4 607 73
          GenBank mRNA BC088427.1 - 3 - 4 537 7124060 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003225.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cldn18

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Process(es)

          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P492543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3605 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P492543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P492543 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P994229 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P621738 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919527 - -
          Affymetrix Rat Gene 1.0 ST Array 10919529 - -
          Affymetrix Rat Gene 1.0 ST Array 10919530 - -
          Affymetrix Rat Gene 1.0 ST Array 10919528 - -
          Affymetrix Rat Gene 1.0 ST Array 10919526 - -
          Affymetrix Rat Gene 1.1 ST Array 10919525 - -
          Rat Genome 230 2.0 Array 1382833_PM_at - -
          Rat Genome 230 2.0 Array 1382833_at - -
          Rat Genome 230 2.0 Array 1383562_PM_at - -
          Rat Genome 230 2.0 Array 1397773_PM_at - -
          Rat Genome 230 2.0 Array 1397773_at - -
          Rat Genome 230 2.0 Array 1383562_at - -
          Affymetrix HT Rat Focus Array 1397773_PM_at - -
          Affymetrix HT Rat Focus Array 1383562_PM_at - -
          Affymetrix HT Rat Focus Array 1383562_at - -
          Affymetrix HT Rat Focus Array 1397773_at - -
          Affymetrix Rat Clariom S Assay PSR0800025753.rn.1 TC0800003225.rn.1 RTA TC
          Rat Expression Set 230 1397773_at - -
          Rat Expression Set 230 1397773_PM_at - -
          Rat Expression Set 230 1383562_at - -
          Rat Expression Set 230 1383562_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919525 - -
          Rat Genome U34 Set rc_AA859001_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21726856 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline