Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447525_g1
          See other SHISA5 GE Assays ›
          Gene Symbol:
          Shisa5
          Gene Name:
          shisa family member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 117781014 - 117795923 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447525_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shisa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006989.1 NP_001006990.1 3 - 4 420 62
          XM_006243760.1 XP_006243822.1 5 - 6 752 62
          XM_006243761.1 XP_006243823.1 5 - 6 749 62
          XM_006243762.2 XP_006243824.1 3 - 4 738 62
          XM_008766511.1 XP_008764733.1 4 - 5 450 62
          GenBank mRNA BC083710.1 - 3 - 4 420 7190608 62
          FQ214071.1 - 3 - 4 396 62
          FQ227338.1 - 3 - 4 425 62
          FQ231597.1 - 5 - 6 736 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001566.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shisa5

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          intrinsic apoptotic signaling pathway by p53 class mediator
          signal transducer activity
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013197 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1030215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1030215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013197 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1030215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2253 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10913492 - -
          Affymetrix Rat Gene 1.0 ST Array 10913491 - -
          Affymetrix Rat Gene 1.0 ST Array 10913493 - -
          Affymetrix Rat Gene 1.0 ST Array 10913494 - -
          Affymetrix Rat Gene 1.0 ST Array 10913495 - -
          Affymetrix Rat Gene 1.1 ST Array 10913487 - -
          Rat Genome 230 2.0 Array 1379891_at - -
          Rat Genome 230 2.0 Array 1388776_PM_at - -
          Rat Genome 230 2.0 Array 1379891_PM_at - -
          Rat Genome 230 2.0 Array 1388776_at - -
          Affymetrix HT Rat Focus Array 1388776_at - -
          Affymetrix HT Rat Focus Array 1388776_PM_at - -
          Affymetrix HT Rat Focus Array 1379891_at - -
          Affymetrix HT Rat Focus Array 1379891_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800012159.rn.1 TC0800001566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800012157.rn.1 TC0800001566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800012158.rn.1 TC0800001566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800012154.rn.1 TC0800001566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800012155.rn.1 TC0800001566.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800012156.rn.1 TC0800001566.rn.1 RTA TC
          Rat Expression Set 230 1388776_PM_at - -
          Rat Expression Set 230 1388776_at - -
          Rat Expression Set 230 1379891_PM_at - -
          Rat Expression Set 230 1379891_at - -
          Affymetrix Rat Gene 1.0 ST Array 10913487 - -
          Rat Genome U34 Set rc_AI169176_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21036693 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367161 - -
          RatRef-12 Expression BeadChip ILMN_68217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline