Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447725_m1
          See other CMTM8 GE Assays ›
          Gene Symbol:
          Cmtm8
          Gene Name:
          CKLF-like MARVEL transmembrane domain containing 8
          Gene Aliases:
          Cklfsf8
          Chromosome Location:
          Chr.8: 122848249 - 122913591 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447725_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cmtm8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198754.2 NP_942049.1 3 - 4 787 76
          XM_006243964.2 XP_006244026.1 3 - 4 493 76
          GenBank mRNA AF538323.1 - 3 - 4 590 76
          BC088094.1 - 3 - 4 787 7325894 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003492.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cmtm8

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P689873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149788 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689873 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P689873 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P689873 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P689873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98038 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16496 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920706 - -
          Affymetrix Rat Gene 1.0 ST Array 10920709 - -
          Affymetrix Rat Gene 1.0 ST Array 10920707 - -
          Affymetrix Rat Gene 1.0 ST Array 10920710 - -
          Affymetrix Rat Gene 1.0 ST Array 10920708 - -
          Affymetrix Rat Gene 1.1 ST Array 10920705 - -
          Rat Genome 230 2.0 Array 1374491_PM_at - -
          Rat Genome 230 2.0 Array 1374491_at - -
          Affymetrix HT Rat Focus Array 1374491_at - -
          Affymetrix HT Rat Focus Array 1374491_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800028422.rn.1 TC0800003492.rn.1 RTA TC
          Rat Expression Set 230 1374491_PM_at - -
          Rat Expression Set 230 1374491_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920705 - -
          Rat Genome U34 Set rc_AA924279_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20862698 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372621 - -
          RatRef-12 Expression BeadChip ILMN_53709 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline