Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01448051_m1
          See other YAP1 GE Assays ›
          Gene Symbol:
          Yap1
          Gene Name:
          yes-associated protein 1
          Gene Aliases:
          YAP65, Yap
          Chromosome Location:
          Chr.8: 6133014 - 6204226 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01448051_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Yap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034002.2 NP_001029174.2 8 - 9 1120 98
          XM_006242488.2 XP_006242550.1 7 - 8 1772 98
          XM_006242489.2 XP_006242551.1 7 - 8 1764 98
          XM_006242490.2 XP_006242552.1 7 - 8 1759 98
          XM_006242491.2 XP_006242553.1 7 - 8 1751 98
          XM_006242492.2 XP_006242554.1 6 - 7 1711 98
          XM_006242493.2 XP_006242555.1 6 - 7 1703 98
          XM_008765893.1 XP_008764115.1 6 - 7 1724 98
          XM_008765894.1 XP_008764116.1 6 - 7 1716 98
          GenBank mRNA DQ186896.2 - 7 - 8 1072 98
          DQ186897.2 - 7 - 8 1084 98
          DQ186898.2 - 8 - 9 1120 98
          DQ376007.1 - 6 - 7 1059 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003670.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Yap1

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          vasculogenesis
          embryonic heart tube morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          cell proliferation
          positive regulation of cell proliferation
          regulation of keratinocyte proliferation
          keratinocyte differentiation
          negative regulation of epithelial cell differentiation
          notochord development
          somatic stem cell population maintenance
          hippo signaling
          regulation of cell proliferation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of organ growth
          paraxial mesoderm development
          lateral mesoderm development
          contact inhibition
          bud elongation involved in lung branching
          lung epithelial cell differentiation
          regulation of canonical Wnt signaling pathway
          cellular response to retinoic acid
          cellular response to gamma radiation
          regulation of stem cell proliferation
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of canonical Wnt signaling pathway
          positive regulation of stem cell population maintenance
          negative regulation of nucleic acid-templated transcription
          negative regulation of stem cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein C-terminus binding
          transcription regulatory region DNA binding
          proline-rich region binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P302952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P302952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P302952 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P302952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914819 - -
          Affymetrix Rat Gene 1.0 ST Array 10914821 - -
          Affymetrix Rat Gene 1.0 ST Array 10914814 - -
          Affymetrix Rat Gene 1.0 ST Array 10914813 - -
          Affymetrix Rat Gene 1.0 ST Array 10914816 - -
          Affymetrix Rat Gene 1.0 ST Array 10914815 - -
          Affymetrix Rat Gene 1.0 ST Array 10914817 - -
          Affymetrix Rat Gene 1.0 ST Array 10914820 - -
          Affymetrix Rat Gene 1.0 ST Array 10914818 - -
          Affymetrix Rat Gene 1.1 ST Array 10914811 - -
          Affymetrix Rat Clariom S Assay PSR0800030169.rn.1 TC0800003670.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800030168.rn.1 TC0800003670.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10914811 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22094743 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370771 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline