Search
Search
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022847.1 | NP_074038.1 | 2 - 3 | 1607 | 53 | |
| GenBank mRNA | HM037361.1 | - | 2159 | 53 | ||
| HM037362.1 | - | 2159 | 53 | |||
| L16922.1 | - | 2 - 3 | 1607 | 53 | ||
Set Membership: |
|