Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021760.1 | NP_068528.1 | 5 - 6 | 752 | 74 | |
XM_008765979.1 | XP_008764201.1 | 5 - 6 | 817 | 74 | ||
GenBank mRNA | AF272661.1 | - | 5 - 6 | 752 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |