Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01448632_m1
          See other SFXN1 GE Assays ›
          Gene Symbol:
          Sfxn1
          Gene Name:
          sideroflexin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 11046588 - 11082161 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01448632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfxn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012213.1 NP_001012213.1 10 - 11 939 80
          XM_006253645.2 XP_006253707.1 10 - 11 1150 80
          XM_008771502.1 XP_008769724.1 10 - 11 911 80
          GenBank mRNA BC078935.1 - 10 - 11 939 7108166 80
          FQ209748.1 - 893 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001359.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfxn1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          erythrocyte differentiation
          ion transmembrane transport
          iron ion homeostasis
          ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138513 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138513 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138513 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37493 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797486 - -
          Affymetrix Rat Gene 1.0 ST Array 10797485 - -
          Affymetrix Rat Gene 1.0 ST Array 10797480 - -
          Affymetrix Rat Gene 1.0 ST Array 10797481 - -
          Affymetrix Rat Gene 1.0 ST Array 10797484 - -
          Affymetrix Rat Gene 1.0 ST Array 10797482 - -
          Affymetrix Rat Gene 1.0 ST Array 10797483 - -
          Affymetrix Rat Gene 1.0 ST Array 10797489 - -
          Affymetrix Rat Gene 1.0 ST Array 10797478 - -
          Affymetrix Rat Gene 1.0 ST Array 10797479 - -
          Affymetrix Rat Gene 1.0 ST Array 10797487 - -
          Affymetrix Rat Gene 1.0 ST Array 10797488 - -
          Affymetrix Rat Gene 1.1 ST Array 10797475 - -
          Rat Genome 230 2.0 Array 1397740_PM_at - -
          Rat Genome 230 2.0 Array 1372715_at - -
          Rat Genome 230 2.0 Array 1372715_PM_at - -
          Rat Genome 230 2.0 Array 1397740_at - -
          Affymetrix HT Rat Focus Array 1397740_at - -
          Affymetrix HT Rat Focus Array 1397740_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009561.rn.1 TC1700001359.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009562.rn.1 TC1700001359.rn.1 RTA TC
          Rat Expression Set 230 1372715_PM_at - -
          Rat Expression Set 230 1372715_at - -
          Rat Expression Set 230 1397740_at - -
          Rat Expression Set 230 1397740_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797475 - -
          Rat Genome U34 Set S70011_at - -
          Rat Genome U34 Set S70011_g_at - -
          Rat Genome U34 Set rc_AA943004_s_at - -
          Rat Genome U34 Set rc_AA819349_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20963572 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376571 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375928 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline