Search
Search
| Gene Symbol: Cltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053835.1 | NP_446287.1 | 1 - 2 | 215 | 135 | |
| XM_006253595.2 | XP_006253657.1 | 1 - 2 | 303 | 135 | ||
| GenBank mRNA | FQ213983.1 | - | 1 - 2 | 305 | 135 | |
| FQ229322.1 | - | 1 - 2 | 234 | 135 | ||
| M15883.1 | - | 1 - 2 | 215 | 135 | ||
| M19262.1 | - | 1 - 2 | 215 | 135 | ||
Set Membership: |
|