Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01448978_m1
          See other TRAM1 GE Assays ›
          Gene Symbol:
          Tram1
          Gene Name:
          translocation associated membrane protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 5055892 - 5064313 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01448978_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tram1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007701.1 NP_001007702.1 1205 86
          GenBank mRNA BC083851.1 - 1205 7123274 86
          FQ223598.1 - 1077 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000050.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tram1

          Gene Ontology Categories:

          Function(s)

          protein transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P552906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P552906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P552906 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P552906 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P107702 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P489120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41076 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867177 - -
          Affymetrix Rat Gene 1.0 ST Array 10867182 - -
          Affymetrix Rat Gene 1.0 ST Array 10867186 - -
          Affymetrix Rat Gene 1.0 ST Array 10867181 - -
          Affymetrix Rat Gene 1.0 ST Array 10867187 - -
          Affymetrix Rat Gene 1.0 ST Array 10867178 - -
          Affymetrix Rat Gene 1.0 ST Array 10867180 - -
          Affymetrix Rat Gene 1.0 ST Array 10867174 - -
          Affymetrix Rat Gene 1.0 ST Array 10867185 - -
          Affymetrix Rat Gene 1.0 ST Array 10867179 - -
          Affymetrix Rat Gene 1.0 ST Array 10867184 - -
          Affymetrix Rat Gene 1.0 ST Array 10867183 - -
          Affymetrix Rat Gene 1.1 ST Array 10867173 - -
          Rat Genome 230 2.0 Array 1390077_at - -
          Rat Genome 230 2.0 Array 1371688_at - -
          Rat Genome 230 2.0 Array 1390077_PM_at - -
          Rat Genome 230 2.0 Array 1371688_PM_at - -
          Affymetrix HT Rat Focus Array 1371688_at - -
          Affymetrix HT Rat Focus Array 1390077_at - -
          Affymetrix HT Rat Focus Array 1371688_PM_at - -
          Affymetrix HT Rat Focus Array 1390077_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500000326.rn.1 TC0500000050.rn.1 RTA TC
          Rat Expression Set 230 1371688_PM_at - -
          Rat Expression Set 230 1371688_at - -
          Rat Expression Set 230 1390077_PM_at - -
          Rat Expression Set 230 1390077_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867173 - -
          Rat Genome U34 Set rc_AA801273_at - -
          Rat Genome U34 Set rc_AI045600_at - -
          Rat Genome U34 Set rc_AA956421_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21937838 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354303 - -
          RatRef-12 Expression BeadChip ILMN_52034 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0