Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449220_m1
          See other POTEA GE Assays ›
          Gene Symbol:
          Potea
          Gene Name:
          POTE ankyrin domain family, member A
          Gene Aliases:
          Ankrd36, Ankrd48, Spergen2
          Chromosome Location:
          Chr.14: 85814809 - 85930536 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449220_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Potea
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008770390.1 XP_008768612.1 13 - 14 1571 89
          XM_008770391.1 XP_008768613.1 13 - 14 1571 89
          XM_008770392.1 XP_008768614.1 12 - 13 1397 89
          XM_008770393.1 XP_008768615.1 8 - 9 2313 89
          GenBank mRNA AB095021.1 - 13 - 14 1440 89
          BC083629.1 - 13 - 14 1548 7134382 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002472.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s)

          spermatogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P287956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33456 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P287956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P287956 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P287956 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P287956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99522 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10773978 - -
          Affymetrix Rat Gene 1.0 ST Array 10773989 - -
          Affymetrix Rat Gene 1.0 ST Array 10773986 - -
          Affymetrix Rat Gene 1.0 ST Array 10773975 - -
          Affymetrix Rat Gene 1.0 ST Array 10773980 - -
          Affymetrix Rat Gene 1.0 ST Array 10773983 - -
          Affymetrix Rat Gene 1.0 ST Array 10773979 - -
          Affymetrix Rat Gene 1.0 ST Array 10773982 - -
          Affymetrix Rat Gene 1.0 ST Array 10773988 - -
          Affymetrix Rat Gene 1.0 ST Array 10773976 - -
          Affymetrix Rat Gene 1.0 ST Array 10773987 - -
          Affymetrix Rat Gene 1.0 ST Array 10773984 - -
          Affymetrix Rat Gene 1.0 ST Array 10773985 - -
          Affymetrix Rat Gene 1.0 ST Array 10773974 - -
          Affymetrix Rat Gene 1.0 ST Array 10773981 - -
          Affymetrix Rat Gene 1.0 ST Array 10773977 - -
          Affymetrix Rat Gene 1.1 ST Array 10773973 - -
          Affymetrix Rat Clariom S Assay PSR1400018408.rn.1 TC1400002472.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018409.rn.1 TC1400002472.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10773973 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21603576 - -
          ABI Rat Genome Survey Microarray 21880517 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359227 - -
          RatRef-12 Expression BeadChip ILMN_59778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline