Search
Search
| Gene Symbol: Sfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191682.1 | NP_001178611.1 | 13 - 14 | 1407 | 62 | |
| XM_006251285.2 | XP_006251347.1 | 14 - 15 | 1806 | 62 | ||
| XM_006251287.2 | XP_006251349.1 | 15 - 16 | 1862 | 62 | ||
| XM_006251288.2 | XP_006251350.1 | 14 - 15 | 1806 | 62 | ||
| XM_006251289.2 | XP_006251351.1 | 14 - 15 | 1806 | 62 | ||
| XM_008770294.1 | XP_008768516.1 | 13 - 14 | 1903 | 62 | ||
| XM_008770295.1 | XP_008768517.1 | 8 - 9 | 1689 | 62 | ||
Set Membership: |
|