Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449537_m1
          See other ERMN GE Assays ›
          Gene Symbol:
          Ermn
          Gene Name:
          ermin, ERM-like protein
          Gene Aliases:
          RGD1308367, juxtanodin
          Chromosome Location:
          Chr.3: 44080350 - 44086006 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449537_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ermn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008311.3 NP_001008312.1 2 - 3 484 82
          GenBank mRNA BC086596.1 - 2 - 3 484 7311836 82
          DQ119821.1 - 412 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003149.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ermn

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of a branching structure
          actin filament organization
          regulation of cell shape
          regulation of cell projection organization
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P779687 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P314110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121283 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P779687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P314110 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P314110 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P779687 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P314110 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P779687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P314110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37376 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19192 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845403 - -
          Affymetrix Rat Gene 1.0 ST Array 10845406 - -
          Affymetrix Rat Gene 1.0 ST Array 10845404 - -
          Affymetrix Rat Gene 1.0 ST Array 10845405 - -
          Affymetrix Rat Gene 1.0 ST Array 10845402 - -
          Affymetrix Rat Gene 1.1 ST Array 10845401 - -
          Rat Genome 230 2.0 Array 1382382_PM_at - -
          Rat Genome 230 2.0 Array 1395447_PM_at - -
          Rat Genome 230 2.0 Array 1382382_at - -
          Rat Genome 230 2.0 Array 1385547_PM_at - -
          Rat Genome 230 2.0 Array 1385547_at - -
          Rat Genome 230 2.0 Array 1395447_at - -
          Affymetrix HT Rat Focus Array 1385547_at - -
          Affymetrix HT Rat Focus Array 1382382_at - -
          Affymetrix HT Rat Focus Array 1385547_PM_at - -
          Affymetrix HT Rat Focus Array 1395447_at - -
          Affymetrix HT Rat Focus Array 1382382_PM_at - -
          Affymetrix HT Rat Focus Array 1395447_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300024315.rn.1 TC0300003149.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300024317.rn.1 TC0300003149.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300024316.rn.1 TC0300003149.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300024314.rn.1 TC0300003149.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300024318.rn.1 TC0300003149.rn.1 RTA TC
          Rat Expression Set 230 1395447_PM_at - -
          Rat Expression Set 230 1382382_at - -
          Rat Expression Set 230 1382382_PM_at - -
          Rat Expression Set 230 1385547_at - -
          Rat Expression Set 230 1385547_PM_at - -
          Rat Expression Set 230 1395447_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845401 - -
          Rat Genome U34 Set rc_AI072720_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22226920 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372576 - -
          RatRef-12 Expression BeadChip ILMN_59575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline