Search
Search

Ccdc148| Gene Symbol: Ccdc148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107732.1 | NP_001101202.1 | 6 - 7 | 787 | 66 | |
| XM_008761833.1 | XP_008760055.1 | 6 - 7 | 873 | 66 | ||
| XM_008761834.1 | XP_008760056.1 | 6 - 7 | 846 | 66 | ||
| XM_008761835.1 | XP_008760057.1 | 6 - 7 | 1370 | 66 | ||
| XM_008761836.1 | XP_008760058.1 | 6 - 7 | 1253 | 66 | ||
| XM_008761837.1 | XP_008760059.1 | 6 - 7 | 873 | 66 | ||
| GenBank mRNA | BC167021.1 | - | 8 - 9 | 1058 | 66 | |
Set Membership: |
|