Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449590_m1
          See other MARCH7 GE Assays ›
          Gene Symbol:
          March7
          Gene Name:
          membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase
          Gene Aliases:
          Axot
          Chromosome Location:
          Chr.3: 46284962 - 46325230 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449590_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: March7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012087.1 NP_001012087.1 7 - 8 2080 135
          XM_008761841.1 XP_008760063.1 7 - 8 2771 135
          XM_008761842.1 XP_008760064.1 6 - 7 1903 135
          XM_008761843.1 XP_008760065.1 6 - 7 1903 135
          XM_008761844.1 XP_008760066.1 7 - 8 2361 135
          GenBank mRNA BC083842.1 - 7 - 8 2080 7114399 135
          FQ222785.1 - 4 - 5 1650 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000657.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene March7

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of tolerance induction
          protein ubiquitination
          negative regulation of T cell proliferation
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P201511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P201511 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P201511 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836403 - -
          Affymetrix Rat Gene 1.0 ST Array 10836399 - -
          Affymetrix Rat Gene 1.0 ST Array 10836395 - -
          Affymetrix Rat Gene 1.0 ST Array 10836402 - -
          Affymetrix Rat Gene 1.0 ST Array 10836396 - -
          Affymetrix Rat Gene 1.0 ST Array 10836398 - -
          Affymetrix Rat Gene 1.0 ST Array 10836401 - -
          Affymetrix Rat Gene 1.0 ST Array 10836400 - -
          Affymetrix Rat Gene 1.0 ST Array 10836397 - -
          Affymetrix Rat Gene 1.1 ST Array 10836394 - -
          Rat Genome 230 2.0 Array 1382125_PM_at - -
          Rat Genome 230 2.0 Array 1373087_PM_at - -
          Rat Genome 230 2.0 Array 1382125_at - -
          Rat Genome 230 2.0 Array 1373087_at - -
          Affymetrix HT Rat Focus Array 1382125_PM_at - -
          Affymetrix HT Rat Focus Array 1382125_at - -
          Affymetrix Rat Clariom S Assay PSR0300005025.rn.1 TC0300000657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005016.rn.1 TC0300000657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005021.rn.1 TC0300000657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005026.rn.1 TC0300000657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005019.rn.1 TC0300000657.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005020.rn.1 TC0300000657.rn.1 RTA TC
          Rat Expression Set 230 1373087_PM_at - -
          Rat Expression Set 230 1373087_at - -
          Rat Expression Set 230 1382125_at - -
          Rat Expression Set 230 1382125_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836394 - -
          Rat Genome U34 Set rc_AA801187_s_at - -
          Rat Genome U34 Set rc_AI171753_at - -
          Rat Genome U34 Set rc_AI175452_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22061051 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376312 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline