Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449785_m1
          See other CYTIP GE Assays ›
          Gene Symbol:
          Cytip
          Gene Name:
          cytohesin 1 interacting protein
          Gene Aliases:
          Pscdbp
          Chromosome Location:
          Chr.3: 44150494 - 44225724 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449785_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cytip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012086.1 NP_001012086.1 7 - 8 658 113
          XM_006234202.2 XP_006234264.1 8 - 9 632 113
          GenBank mRNA BC088207.1 - 7 - 8 658 7381695 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003150.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cytip

          Gene Ontology Categories:

          Function(s)

          regulation of cell adhesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480573 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480573 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23262 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75970 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845409 - -
          Affymetrix Rat Gene 1.0 ST Array 10845408 - -
          Affymetrix Rat Gene 1.0 ST Array 10845415 - -
          Affymetrix Rat Gene 1.0 ST Array 10845410 - -
          Affymetrix Rat Gene 1.0 ST Array 10845413 - -
          Affymetrix Rat Gene 1.0 ST Array 10845412 - -
          Affymetrix Rat Gene 1.0 ST Array 10845414 - -
          Affymetrix Rat Gene 1.0 ST Array 10845411 - -
          Affymetrix Rat Gene 1.1 ST Array 10845407 - -
          Affymetrix Rat Clariom S Assay PSR0300024328.rn.1 TC0300003150.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10845407 - -
          Rat Genome U34 Set rc_AI012428_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22069607 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline