Search
Search
| Gene Symbol: Wdr38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761808.1 | XP_008760030.1 | 7 - 8 | 903 | 82 | |
| XM_575125.5 | XP_575125.6 | 7 - 8 | 863 | 82 | ||
| GenBank mRNA | FQ213095.1 | - | 7 - 8 | 857 | 82 | |
Set Membership: |
|