Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450155_m1
          See other AGPAT9 GE Assays ›
          Gene Symbol:
          Agpat9
          Gene Name:
          1-acylglycerol-3-phosphate O-acyltransferase 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 10343290 - 10396561 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450155_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Agpat9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025670.1 NP_001020841.1 9 - 10 1292 70
          XM_008770019.1 XP_008768241.1 10 - 11 1289 70
          XM_008770020.1 XP_008768242.1 10 - 11 1718 70
          XM_008770021.1 XP_008768243.1 10 - 11 1417 70
          GenBank mRNA BC097362.1 - 9 - 10 1292 7442656 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001371.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Agpat9

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          CDP-diacylglycerol biosynthetic process
          triglyceride biosynthetic process
          regulation of TOR signaling
          1-acylglycerol-3-phosphate O-acyltransferase activity
          glycerol-3-phosphate O-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P765736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P765736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P765736 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P765736 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P234071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775525 - -
          Affymetrix Rat Gene 1.0 ST Array 10775523 - -
          Affymetrix Rat Gene 1.0 ST Array 10775528 - -
          Affymetrix Rat Gene 1.0 ST Array 10775521 - -
          Affymetrix Rat Gene 1.0 ST Array 10775526 - -
          Affymetrix Rat Gene 1.0 ST Array 10775524 - -
          Affymetrix Rat Gene 1.0 ST Array 10775520 - -
          Affymetrix Rat Gene 1.0 ST Array 10775531 - -
          Affymetrix Rat Gene 1.0 ST Array 10775522 - -
          Affymetrix Rat Gene 1.0 ST Array 10775527 - -
          Affymetrix Rat Gene 1.0 ST Array 10775530 - -
          Affymetrix Rat Gene 1.0 ST Array 10775529 - -
          Affymetrix Rat Gene 1.1 ST Array 10775519 - -
          Affymetrix Rat Clariom S Assay PSR1400010223.rn.1 TC1400001371.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10775519 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21987964 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline