Search
Search

Rasgef1b| Gene Symbol: Rasgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728083.4 | XP_002728129.2 | 8 - 9 | 1098 | 79 | |
| XM_008770077.1 | XP_008768299.1 | 8 - 9 | 1095 | 79 | ||
Set Membership: |
|