Search
Search
| Gene Symbol: Cops4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004275.1 | NP_001004275.1 | 4 - 5 | 448 | 90 | |
| XM_006250667.2 | XP_006250729.1 | 3 - 4 | 358 | 90 | ||
| XM_006250668.2 | XP_006250730.1 | 4 - 5 | 494 | 90 | ||
| GenBank mRNA | BC079384.1 | - | 4 - 5 | 448 | 7111425 | 90 |
Set Membership: |
|