Search
Search

RGD1561145| Gene Symbol: RGD1561145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134605.2 | NP_001128077.2 | 2 - 3 | 137 | 68 | |
| XM_008771144.1 | XP_008769366.1 | 2 - 3 | 293 | 68 | ||
| GenBank mRNA | CB315636.1 | - | 2 - 3 | 140 | 68 | |
Set Membership: |
|