Search
Search

Ankrd28| Gene Symbol: Ankrd28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252707.2 | XP_006252769.1 | 15 - 16 | 1984 | 65 | |
| XM_006252708.2 | XP_006252770.1 | 14 - 15 | 1913 | 65 | ||
| XM_008771058.1 | XP_008769280.1 | 17 - 18 | 7792 | 65 | ||
Set Membership: |
|