Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450394_m1
          See other DNAH1 GE Assays ›
          Gene Symbol:
          Dnah1
          Gene Name:
          dynein, axonemal, heavy chain 1
          Gene Aliases:
          Dnahc1, RGD1311110
          Chromosome Location:
          Chr.16: 7345131 - 7407009 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450394_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnah1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033655.2 NP_001028827.2 56 - 57 9023 69
          XM_008770977.1 XP_008769199.1 56 - 57 9081 69
          XM_008770978.1 XP_008769200.1 56 - 57 9081 69
          XM_008770979.1 XP_008769201.1 56 - 57 9081 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001175.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnah1

          Panther Classification:

          Molecular Function -

          cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cilium movement
          epithelial cilium movement
          metabolic process
          sperm motility
          cilium movement involved in cell motility
          microtubule motor activity
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5813 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158738 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158738 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16210 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1003917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25089 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790133 - -
          Affymetrix Rat Gene 1.0 ST Array 10790205 - -
          Affymetrix Rat Gene 1.0 ST Array 10790141 - -
          Affymetrix Rat Gene 1.0 ST Array 10790204 - -
          Affymetrix Rat Gene 1.0 ST Array 10790138 - -
          Affymetrix Rat Gene 1.0 ST Array 10790199 - -
          Affymetrix Rat Gene 1.0 ST Array 10790149 - -
          Affymetrix Rat Gene 1.0 ST Array 10790168 - -
          Affymetrix Rat Gene 1.0 ST Array 10790180 - -
          Affymetrix Rat Gene 1.0 ST Array 10790207 - -
          Affymetrix Rat Gene 1.0 ST Array 10790148 - -
          Affymetrix Rat Gene 1.0 ST Array 10790146 - -
          Affymetrix Rat Gene 1.0 ST Array 10790134 - -
          Affymetrix Rat Gene 1.0 ST Array 10790179 - -
          Affymetrix Rat Gene 1.0 ST Array 10790195 - -
          Affymetrix Rat Gene 1.0 ST Array 10790192 - -
          Affymetrix Rat Gene 1.0 ST Array 10790210 - -
          Affymetrix Rat Gene 1.0 ST Array 10790188 - -
          Affymetrix Rat Gene 1.0 ST Array 10790213 - -
          Affymetrix Rat Gene 1.0 ST Array 10790136 - -
          Affymetrix Rat Gene 1.0 ST Array 10790211 - -
          Affymetrix Rat Gene 1.0 ST Array 10790176 - -
          Affymetrix Rat Gene 1.0 ST Array 10790169 - -
          Affymetrix Rat Gene 1.0 ST Array 10790152 - -
          Affymetrix Rat Gene 1.0 ST Array 10790181 - -
          Affymetrix Rat Gene 1.0 ST Array 10790212 - -
          Affymetrix Rat Gene 1.0 ST Array 10790144 - -
          Affymetrix Rat Gene 1.0 ST Array 10790193 - -
          Affymetrix Rat Gene 1.0 ST Array 10790162 - -
          Affymetrix Rat Gene 1.0 ST Array 10790142 - -
          Affymetrix Rat Gene 1.0 ST Array 10790170 - -
          Affymetrix Rat Gene 1.0 ST Array 10790160 - -
          Affymetrix Rat Gene 1.0 ST Array 10790189 - -
          Affymetrix Rat Gene 1.0 ST Array 10790178 - -
          Affymetrix Rat Gene 1.0 ST Array 10790166 - -
          Affymetrix Rat Gene 1.0 ST Array 10790177 - -
          Affymetrix Rat Gene 1.0 ST Array 10790174 - -
          Affymetrix Rat Gene 1.0 ST Array 10790165 - -
          Affymetrix Rat Gene 1.0 ST Array 10790171 - -
          Affymetrix Rat Gene 1.0 ST Array 10790197 - -
          Affymetrix Rat Gene 1.0 ST Array 10790172 - -
          Affymetrix Rat Gene 1.0 ST Array 10790145 - -
          Affymetrix Rat Gene 1.0 ST Array 10790196 - -
          Affymetrix Rat Gene 1.0 ST Array 10790200 - -
          Affymetrix Rat Gene 1.0 ST Array 10790163 - -
          Affymetrix Rat Gene 1.0 ST Array 10790143 - -
          Affymetrix Rat Gene 1.0 ST Array 10790184 - -
          Affymetrix Rat Gene 1.0 ST Array 10790183 - -
          Affymetrix Rat Gene 1.0 ST Array 10790158 - -
          Affymetrix Rat Gene 1.0 ST Array 10790173 - -
          Affymetrix Rat Gene 1.0 ST Array 10790206 - -
          Affymetrix Rat Gene 1.0 ST Array 10790208 - -
          Affymetrix Rat Gene 1.0 ST Array 10790150 - -
          Affymetrix Rat Gene 1.0 ST Array 10790135 - -
          Affymetrix Rat Gene 1.0 ST Array 10790191 - -
          Affymetrix Rat Gene 1.0 ST Array 10790156 - -
          Affymetrix Rat Gene 1.0 ST Array 10790140 - -
          Affymetrix Rat Gene 1.0 ST Array 10790154 - -
          Affymetrix Rat Gene 1.0 ST Array 10790209 - -
          Affymetrix Rat Gene 1.0 ST Array 10790137 - -
          Affymetrix Rat Gene 1.0 ST Array 10790185 - -
          Affymetrix Rat Gene 1.0 ST Array 10790157 - -
          Affymetrix Rat Gene 1.0 ST Array 10790147 - -
          Affymetrix Rat Gene 1.0 ST Array 10790164 - -
          Affymetrix Rat Gene 1.0 ST Array 10790161 - -
          Affymetrix Rat Gene 1.0 ST Array 10790151 - -
          Affymetrix Rat Gene 1.0 ST Array 10790155 - -
          Affymetrix Rat Gene 1.0 ST Array 10790194 - -
          Affymetrix Rat Gene 1.0 ST Array 10790175 - -
          Affymetrix Rat Gene 1.0 ST Array 10790201 - -
          Affymetrix Rat Gene 1.0 ST Array 10790139 - -
          Affymetrix Rat Gene 1.0 ST Array 10790190 - -
          Affymetrix Rat Gene 1.0 ST Array 10790182 - -
          Affymetrix Rat Gene 1.0 ST Array 10790159 - -
          Affymetrix Rat Gene 1.0 ST Array 10790202 - -
          Affymetrix Rat Gene 1.0 ST Array 10790187 - -
          Affymetrix Rat Gene 1.0 ST Array 10790167 - -
          Affymetrix Rat Gene 1.0 ST Array 10790186 - -
          Affymetrix Rat Gene 1.1 ST Array 10790132 - -
          Rat Genome 230 2.0 Array 1381039_at - -
          Rat Genome 230 2.0 Array 1381039_PM_at - -
          Affymetrix HT Rat Focus Array 1381039_at - -
          Affymetrix HT Rat Focus Array 1381039_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600008885.rn.1 TC1600001175.rn.1 RTA TC
          Rat Expression Set 230 1381039_PM_at - -
          Rat Expression Set 230 1381039_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790132 - -
          Rat Genome U34 Set D26492_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21024350 - -
          ABI Rat Genome Survey Microarray 21290010 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367533 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline