Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450559_m1
          See other SIRT5 GE Assays ›
          Gene Symbol:
          Sirt5
          Gene Name:
          sirtuin 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 23986796 - 24013854 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450559_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sirt5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004256.1 NP_001004256.1 6 - 7 790 66
          XM_006253801.1 XP_006253863.1 7 - 8 974 66
          XM_006253802.1 XP_006253864.1 7 - 8 852 66
          XM_006253803.2 XP_006253865.1 7 - 8 846 66
          XM_006253804.2 XP_006253866.1 7 - 8 797 66
          GenBank mRNA BC078958.1 - 6 - 7 790 7110841 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000395.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sirt5

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deacetylation
          negative regulation of cardiac muscle cell apoptotic process
          response to nutrient levels
          peptidyl-lysine demalonylation
          protein desuccinylation
          peptidyl-lysine desuccinylation
          negative regulation of reactive oxygen species metabolic process
          molecular_function
          zinc ion binding
          protein-malonyllysine demalonylase activity
          protein-succinyllysine desuccinylase activity
          NAD+ binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152199 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089915 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1060459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44295 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797832 - -
          Affymetrix Rat Gene 1.0 ST Array 10797834 - -
          Affymetrix Rat Gene 1.0 ST Array 10797828 - -
          Affymetrix Rat Gene 1.0 ST Array 10797830 - -
          Affymetrix Rat Gene 1.0 ST Array 10797831 - -
          Affymetrix Rat Gene 1.0 ST Array 10797833 - -
          Affymetrix Rat Gene 1.0 ST Array 10797835 - -
          Affymetrix Rat Gene 1.0 ST Array 10797829 - -
          Affymetrix Rat Gene 1.1 ST Array 10797827 - -
          Rat Genome 230 2.0 Array 1388925_PM_at - -
          Rat Genome 230 2.0 Array 1388925_at - -
          Affymetrix HT Rat Focus Array 1388925_PM_at - -
          Affymetrix HT Rat Focus Array 1388925_at - -
          Affymetrix Rat Clariom S Assay PSR1700002941.rn.1 TC1700000395.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700002940.rn.1 TC1700000395.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700002942.rn.1 TC1700000395.rn.1 RTA TC
          Rat Expression Set 230 1388925_at - -
          Rat Expression Set 230 1388925_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797827 - -
          Rat Genome U34 Set rc_AI171547_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21782627 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349757 - -
          RatRef-12 Expression BeadChip ILMN_53614 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline