Search
Search
| Gene Symbol: Iars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100572.1 | NP_001094042.1 | 32 - 33 | 3761 | 59 | |
| XM_006253706.2 | XP_006253768.1 | 32 - 33 | 3651 | 59 | ||
| XM_008771533.1 | XP_008769755.1 | 33 - 34 | 3859 | 59 | ||
| GenBank mRNA | BC091369.1 | - | 5 - 6 | 623 | 59 | |
Set Membership: |
|