Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450778_g1
          See other IRF7 GE Assays ›
          Gene Symbol:
          Irf7
          Gene Name:
          interferon regulatory factor 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 214249388 - 214252909 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450778_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033691.1 NP_001028863.1 8 - 9 1619 66
          XM_006230540.1 XP_006230602.1 9 - 10 1718 66
          XM_006230541.1 XP_006230603.1 9 - 10 1622 66
          GenBank mRNA BC104686.1 - 8 - 9 1619 7442863 66
          FQ232130.1 - 7 - 8 1074 66
          FQ232859.1 - 6 - 7 911 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006940.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irf7

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of adaptive immune response
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          immunoglobulin mediated immune response
          interferon-alpha production
          interferon-beta production
          positive regulation of interferon-alpha production
          regulation of MyD88-dependent toll-like receptor signaling pathway
          regulation of MyD88-independent toll-like receptor signaling pathway
          type I interferon biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of type I interferon-mediated signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1039994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039994 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1039994 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039994 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726760 - -
          Affymetrix Rat Gene 1.0 ST Array 10726761 - -
          Affymetrix Rat Gene 1.0 ST Array 10726768 - -
          Affymetrix Rat Gene 1.0 ST Array 10726764 - -
          Affymetrix Rat Gene 1.0 ST Array 10726765 - -
          Affymetrix Rat Gene 1.0 ST Array 10726763 - -
          Affymetrix Rat Gene 1.0 ST Array 10726767 - -
          Affymetrix Rat Gene 1.0 ST Array 10726766 - -
          Affymetrix Rat Gene 1.0 ST Array 10726762 - -
          Affymetrix Rat Gene 1.0 ST Array 10726759 - -
          Affymetrix Rat Gene 1.1 ST Array 10726758 - -
          Rat Genome 230 2.0 Array 1383564_PM_at - -
          Rat Genome 230 2.0 Array 1383564_at - -
          Affymetrix HT Rat Focus Array 1383564_PM_at - -
          Affymetrix HT Rat Focus Array 1383564_at - -
          Affymetrix Rat Clariom S Assay PSR0100054705.rn.1 TC0100006940.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054707.rn.1 TC0100006940.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100054706.rn.1 TC0100006940.rn.1 RTA TC
          Rat Expression Set 230 1383564_at - -
          Rat Expression Set 230 1383564_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726758 - -
          Rat Genome U34 Set rc_AA799861_g_at - -
          Rat Genome U34 Set rc_AA799861_at - -
          Rat Genome U34 Set rc_AA965186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21945611 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline