Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451209_g1
          See other UBXN1 GE Assays ›
          Gene Symbol:
          Ubxn1
          Gene Name:
          UBX domain protein 1
          Gene Aliases:
          2B28, RGD1309471
          Chromosome Location:
          Chr.1: 225067909 - 225071946 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451209_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubxn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034829.1 NP_001030001.1 6 - 7 541 111
          XM_006230975.1 XP_006231037.1 7 - 8 718 111
          GenBank mRNA AJ277751.1 - 6 - 7 541 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubxn1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein ubiquitination
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein binding
          polyubiquitin binding
          ubiquitin protein ligase binding
          ubiquitin binding
          ATPase binding
          K6-linked polyubiquitin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P388572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P388572 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P388572 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713693 - -
          Affymetrix Rat Gene 1.0 ST Array 10713691 - -
          Affymetrix Rat Gene 1.0 ST Array 10713694 - -
          Affymetrix Rat Gene 1.0 ST Array 10713688 - -
          Affymetrix Rat Gene 1.0 ST Array 10713689 - -
          Affymetrix Rat Gene 1.0 ST Array 10713690 - -
          Affymetrix Rat Gene 1.0 ST Array 10713692 - -
          Affymetrix Rat Gene 1.1 ST Array 10713685 - -
          Rat Genome 230 2.0 Array 1373299_at - -
          Rat Genome 230 2.0 Array 1373299_PM_at - -
          Affymetrix HT Rat Focus Array 1373299_PM_at - -
          Affymetrix HT Rat Focus Array 1373299_at - -
          Affymetrix Rat Clariom S Assay PSR0100025977.rn.1 TC0100003158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025978.rn.1 TC0100003158.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025979.rn.1 TC0100003158.rn.1 RTA TC
          Rat Expression Set 230 1373299_at - -
          Rat Expression Set 230 1373299_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713685 - -
          Rat Genome U34 Set rc_AA800039_s_at - -
          Rat Genome U34 Set rc_AI012216_at - -
          Rat Genome U34 Set AA801051_at - -
          Rat Genome U34 Set rc_AI168974_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21624589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline